Skip to Content
Merck
All Photos(1)

Key Documents

C6164

Sigma-Aldrich

Cell Freezing Medium-DMSO 1×

sterile-filtered, suitable for cell culture

Synonym(s):

cryopreservation medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

sterility

sterile-filtered

Quality Level

form

liquid

technique(s)

cell culture | mammalian: suitable
cryopreservation: suitable

impurities

endotoxin, tested

shipped in

dry ice

storage temp.

−20°C

General description

Cell freezing or cryopreservation of cell culture is a routine process. It is essential to maintain the cellular integrity during storage. Majorly used cryoprotectants include glycerol, ethylene glycol and dimethyl sulfoxide(DMSO).

Application

Cell Freezing Medium-DMSO 1× has been used in the cryopreservation of human embryonic kidney HEK293 CNR2 cells, blood mononuclear cells and prostate cancer cells.

Other Notes

Complete ready-to-use medium. Proprietary formulation with Minimum Essential Medium, dimethyl sulfoxide, calf serum and fetal bovine serum.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonia Patsialou et al.
Cancer research, 69(24), 9498-9506 (2009-11-26)
Colony-stimulating factor-1 (CSF-1) and its receptor (CSF-1R) have been implicated in the pathogenesis and progression of various types of cancer, including breast cancer. This is based on high levels of circulating CSF-1 in patient sera with aggressive disease and increased
Directional freezing for the cryopreservation of adherent mammalian cells on a substrate
Bahari L, et al.
Testing, 13(2), e0192265-e0192265 (2018)
Safety and immunogenicity of oral microencapsulated enterotoxigenic Escherichia coli surface antigen 6 (CS6) with and without LTR192G: a randomized clinical trial
Lapa JA, et al.
Clinical and Vaccine Immunology : CVI (2008)
Variations of components of the plasminogen activation system with the cell cycle in benign prostate tissue and prostate cancer
Plas E, et al.
Cytometry, 46(3), 184-189 (2001)
Zhenming Yu et al.
PloS one, 6(3), e17951-e17951 (2011-04-12)
Amyotrophic lateral sclerosis (ALS) is a devastating, rapidly progressive disease leading to paralysis and death. Recently, intermediate length polyglutamine (polyQ) repeats of 27-33 in ATAXIN-2 (ATXN2), encoding the ATXN2 protein, were found to increase risk for ALS. In ATXN2, polyQ

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service