Skip to Content
Merck
All Photos(1)

Key Documents

D5030

Sigma-Aldrich

Dulbecco′s Modified Eagle′s Medium

Without glucose, L-glutamine, phenol red, sodium pyruvate and sodium bicarbonate, powder, suitable for cell culture

Synonym(s):

DME, DMEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

components

phenol red: no
NaHCO3: no
L-glutamine: no
HEPES: no
glucose: no
sodium pyruvate: no

shipped in

ambient

storage temp.

2-8°C

General description

Dulbecco′s Modified Eagle′s Medium (DME) is a modification of Basal Medium Eagle (BME) that has been formulated with a 4-fold higher concentration of amino acids and vitamins. It includes additional supplementary components. The DME formula, first reported for culturing embryonic mouse cells, contained 1,000 mg/L of glucose. Modifying the medium with 4,500 mg/L glucose is optimal for culturing certain cell types.
The most basic formulation offered. This formulation is used by investigators who want to start with the essential components of DME, and have the flexibility to optimize the formula for their own application.

Application

Dulbecco′s Modified Eagle′s Medium has been used:
  • to culture the isolated flexor digitorum brevis (FDB) fibers to measure exogenous fatty acid (FA) utilization as part of oxygen consumption rate (OCR) measurements
  • to culture the isolated tibialis anterior (TA) muscle fibers for lactate measurements
  • to culture the human bone marrow mesenchymal stem cells for osteogenic differentiation to prepare a low-glucose medium to culture human hepatocellular carcinoma HepG2 cells

Quantity

Formulated to contain 8.3 grams of powder per liter of medium.

Reconstitution

Supplement with 1.0 g/L glucose, 0.584 g/L L-glutamine, 3.7 g/L sodium bicarbonate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abhinav Joshi et al.
BMC biology, 18(1), 10-10 (2020-01-29)
The molecular chaperone TRAP1, the mitochondrial isoform of cytosolic HSP90, remains poorly understood with respect to its pivotal role in the regulation of mitochondrial metabolism. Most studies have found it to be an inhibitor of mitochondrial oxidative phosphorylation (OXPHOS) and
Eunyong Ahn et al.
Molecular systems biology, 13(11), 953-953 (2017-11-08)
Cellular metabolic demands change throughout the cell cycle. Nevertheless, a characterization of how metabolic fluxes adapt to the changing demands throughout the cell cycle is lacking. Here, we developed a temporal-fluxomics approach to derive a comprehensive and quantitative view of
Nataly Stylianou et al.
Oncogene, 38(7), 913-934 (2018-09-09)
The propensity of cancer cells to transition between epithelial and mesenchymal phenotypic states via the epithelial-mesenchymal transition (EMT) program can regulate metastatic processes, cancer progression, and treatment resistance. Transcriptional investigations using reversible models of EMT, revealed the mesenchymal-to-epithelial reverting transition
Charlotte Petersen et al.
Scientific reports, 7(1), 13101-13101 (2017-10-14)
Adipose tissue takes up glucose and releases lactate, thereby contributing significantly to systemic glucose and lactate homeostasis. This implies the necessity of upregulation of net acid and lactate flux capacity during adipocyte differentiation and function. However, the regulation of lactate-
Zhongjie Fu et al.
EMBO molecular medicine, 10(1), 76-90 (2017-11-29)
The neural cells and factors determining normal vascular growth are not well defined even though vision-threatening neovessel growth, a major cause of blindness in retinopathy of prematurity (ROP) (and diabetic retinopathy), is driven by delayed normal vascular growth. We here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service