Skip to Content
Merck
All Photos(1)

Key Documents

C2874

Sigma-Aldrich

CryoStor® cell cryopreservation media

CS10

Synonym(s):

cell freezing medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable
cryopreservation: suitable

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

The CryoStor CS2, CS5, and CS10 family of preservation solutions represents the next generation of cryopreservation media. Designed to prepare and preserve cells in ultra low temperature environments (–80 to –196 °C), CryoStor media provide a safe, protective environment for cells and tissues during thefreezing, storage, and thawing process. Through modulating the cellular biochemical response to the cryopreservation process, these media provide enhanced cell viability and functionality, while eliminating the need to include serum, proteins, or high levels of cytotoxic agents.

CryoStor CS2, CS5, and CS10 are a series of cell specific, optimized preservation media, uniquely formulated to address the molecular biological aspects of cells during the cryopreservation process; thereby, directly reducing the level of Cryopreservation-Induced Delayed-Onset Cell Death and improving post-thaw cell viability and function.

These media are recommended for the preservation of stem cells, hepatocytes, tissue samples, and other extremely sensitive cell types.

Application

CryoStor, a series of cell-specific, optimized preservation media, is uniquely formulated to address the molecularbiological aspects of cells during the cryopreservation process thereby directly reducing the level of Cryopreservation-Induced Delayed-Onset Cell Death and improving post-thaw cell viability and function.
CryoStor® cell cryopreservation media has been used in:
  • the preservation of T cells
  • the preservation of human induced pluripotent stem cell-derived cardiomyocytes (hiPSC-CMs)
  • hiPSC-derived cardiac progenitors (hiPSC-CPs)
  • as a component of cryoprotective solution composed of bovine albumin

Other Notes

Formulation contains 10% DMSO.

Legal Information

CryoStor is a registered trademark of BioLife Solutions, Inc.

comparable product

Product No.
Description
Pricing

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Physical events occurring during the cryopreservation of immortalized human T cells
Meneghel J, et al.
PLoS ONE, 14(5), e0217304-e0217304 (2019)
Susie Barbeau et al.
Stem cell research, 49, 102106-102106 (2020-12-30)
Congenital myasthenic syndromes (CMS) are a class of inherited disorders affecting the neuromuscular junction, a synapse whose activity is essential for movement. CMS with acetylcholinesterase (AChE) deficiency are caused by mutations in COLQ, a collagen that anchors AChE in the
Huyen Thi Lam Nguyen et al.
STAR protocols, 1(2) (2020-10-13)
Tumor organoids are promising tools for cancer biology investigations and preclinical drug screenings because they are often representative of the histology and drug responses of patients. Here, we introduce a facile protocol to overcome technical limitations by generating patient-derived tumor
In vivo maturation of human induced pluripotent stem cell-derived cardiomyocytes in neonatal and adult rat hearts
Kadota S, et al.
Stem Cell Reports, 8(2), 278-289 (2017)
The impact of varying cooling and thawing rates on the quality of cryopreserved human peripheral blood T cells
Baboo J, et al.
Scientific reports, 9(1), 3417-3417 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service