Skip to Content
Merck
All Photos(1)

Key Documents

G5154

Sigma-Aldrich

Glasgow Minimum Essential Medium

With sodium bicarbonate, without ʟ-glutamine, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

BHK medium, GMEM, Glasgow′s MEM

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Product Name

Glasgow Minimum Essential Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

L-glutamine: no
phenol red: 0.0213 g/L
NaHCO3: 2.75 g/L
glucose: 4.5 g/L (Dextro)

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

For use with adherent kidney cell lines such as baby hamster kidney cells (BHK).
Glasgow Minimum Essential Medium has been developed by modifying the Eagle′s BME medium. This medium can be used to study the genetic factors affecting cell competence.

Application

Glasgow Minimum Essential Medium has been used to culture:
  • Hoxb7- green fluorescence protein (GFP) mouse embryonic stem (ES) cell line for the induction of ureteric bud differentiation
  • murine E14 Tg2a embryonic stem (mES) cells
  • transduced Chinese hamster ovary (CHO) cells for the expression and production of Siglec-1-Fc fusion proteins

Reconstitution

Supplement with 0.292 g/L L-glutamine and Tryptose phosphate broth solution (T 8159) at 100 ml/L of medium.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco D'Addio et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 35(11), e22017-e22017 (2021-10-27)
Cellular interactions between endothelial cells and macrophages regulate macrophage localization and phenotype, but the mechanisms underlying these interactions are poorly understood. Here we explored the role of sialoglycans on lymphatic endothelial cells (LEC) in interactions with macrophage-expressed Siglec-1 (CD169). Lectin-binding
Marieke Aarts et al.
Genes & development, 31(20), 2085-2098 (2017-11-16)
Expression of the transcription factors OCT4, SOX2, KLF4, and cMYC (OSKM) reprograms somatic cells into induced pluripotent stem cells (iPSCs). Reprogramming is a slow and inefficient process, suggesting the presence of safeguarding mechanisms that counteract cell fate conversion. One such
Sacha Robert et al.
International journal of molecular sciences, 22(19) (2021-10-14)
Differentiation of pluripotent stem cells to cardiomyocytes is influenced by culture conditions including the extracellular matrices or similar synthetic scaffolds on which they are grown. However, the molecular mechanisms that link the scaffold with differentiation outcomes are not fully known.
Cinzia Caprio et al.
Disease models & mechanisms (2021-02-21)
The Ezh2 gene encodes a histone methyltransferase of the Polycomb Repressive Complex 2 that methylates histone H3 lysine 27. In this work we asked whether EZH2 has a role in the development of the pharyngeal apparatus and whether it regulates
Edward C Hsiao et al.
PloS one, 3(7), e2532-e2532 (2008-07-04)
Fluorescent reporters are useful for assaying gene expression in living cells and for identifying and isolating pure cell populations from heterogeneous cultures, including embryonic stem (ES) cells. Multiple fluorophores and genetic selection markers exist; however, a system for creating reporter

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service