Skip to Content
Merck
All Photos(1)

Documents

D5648

Sigma-Aldrich

DMEM - high glucose

With ʟ-glutamine, without sodium bicarbonate and sodium pyruvate, powder, suitable for cell culture

Synonym(s):

DME, Dulbecco′s Modified Eagle′s Medium - high glucose, DMEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

product name

Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture

form

powder

technique(s)

cell culture | mammalian: suitable

components

phenol red: yes
L-glutamine: yes
NaHCO3: no
HEPES: no
glucose: high
sodium pyruvate: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Dulbecco′s Modified Eagle′s Medium - high glucose has been used as the culture medium for various cell types including the human cervical endocervical epithelial End1/E6E7 cells, human adenocarcinoma epithelial cells (HeLa), human alveolar adenocarcinoma epithelial cells (A549), human osteosarcoma epithelial cells, human diploid cell line , primary hepatocytes and synovial tissue
Dulbecco′s Modified Eagle′s Medium (DMEM) is a modification of Basal Medium Eagle (BME) that contains four-fold concentrations of the amino acids and vitamins. The original formulation contained 1000 mg/L of glucose and was used to culture embryonic mouse cells. Since then, it has been modified in several ways to support primary cultures of mouse and chicken cells, as well as a variety of normal and transformed cells. Each of these media offers a different combination of L-glutamine and sodium pyruvate. Additionally, the glucose levels have been raised to 4500 mg/L, contributing to the name "DMEM/High".

Quantity

Formulated to contain 13.4 grams of powder per liter of medium.

Reconstitution

Supplement with 3.7 g/L sodium bicarbonate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisa Levine et al.
Scientific reports, 10(1), 6398-6398 (2020-04-15)
Preeclampsia is the most common placental pathology in pregnant females, with increased morbidity and mortality incurred on the mother and the fetus. There is a need for improved biomarkers for diagnosis and monitoring of this condition. Placental syncytiotrophoblasts at the
Enrico Luchinat et al.
Analytical chemistry, 92(14), 9997-10006 (2020-06-20)
In-cell NMR can investigate protein conformational changes at atomic resolution, such as those changes induced by drug binding or chemical modifications, directly in living human cells, and therefore has great potential in the context of drug development as it can
Veronika Paluchova et al.
Diabetes, 69(3), 300-312 (2019-12-07)
Branched esters of palmitic acid and hydroxystearic acid (PAHSA) are anti-inflammatory and antidiabetic lipokines that connect glucose and lipid metabolism. We aimed to characterize involvement of the 5-PAHSA regioisomer in the adaptive metabolic response of white adipose tissue (WAT) to
Jelka Gabrilovac et al.
Immunobiology, 222(2), 327-342 (2016-09-30)
Dipeptidyl peptidase 9 (DPP9) is a relatively new member of the DPPIV family of prolyl dipeptidases which is ubiquitously expressed. Its role in regulation of immune responses and proliferation of epithelial carcinoma cells was reported. There is no data on
Huan Lin et al.
Nature communications, 9(1), 1875-1875 (2018-05-16)
It has been generally thought that tRNA modifications are stable and static, and their frequencies are rarely regulated. N6-threonylcarbamoyladenosine (t6A) occurs at position 37 of five mitochondrial (mt-)tRNA species. We show that YRDC and OSGEPL1 are responsible for t6A37 formation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service