Skip to Content
Merck
All Photos(1)

Key Documents

650552

Sigma-Aldrich

Hexane

≥95%, HPLC Plus, for residue analysis, suitable for gas chromatography (GC) and HPLC, suitable for EPA 1613, EPA ACB B21-02, EPA OTM-45, and EPA ACB B23-05b

Synonym(s):

n-Hexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH3
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.06
grade:
HPLC Plus
for residue analysis
technique(s):
HPLC: suitable
gas chromatography (GC): suitable
application(s):
food and beverages
bp:
69 °C (lit.)
vapor pressure:
256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

Product Name

Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%

grade

HPLC Plus
for residue analysis

Agency

suitable for EPA 1613
suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b
suitable for EPA OTM-45

vapor density

~3 (vs air)

vapor pressure

256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

Assay

≥95%

form

liquid

autoignition temp.

453 °F

expl. lim.

7.7 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤ 1.0 ppb Fluorescence (quinine) at 365 nm
<0.01% water

evapn. residue

<0.0001%

halogenated residue

<10 ng/L (as heptachlor epoxide)

refractive index

n20/D 1.375 (lit.)

pH

7

bp

69 °C (lit.)

mp

−95 °C (lit.)

solubility

ethanol: soluble(lit.)

density

0.659 g/mL at 25 °C (lit.)

UV absorption

λ: 195 nm Amax: 1.00
λ: 200 nm Amax: 0.50
λ: 210 nm Amax: 0.20
λ: 220 nm Amax: 0.05
λ: 250 nm Amax: 0.01
λ: 280-400 nm Amax: 0.005

suitability

suitable for residue analysis

application(s)

food and beverages

SMILES string

CCCCCC

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexane (n-hexane) is an alkane, that can be obtained from petroleum by distillation. It can also be produced by electrolyzing the potassium butyrate solution. It is widely employed organic solvent for the dissolution and isolation of fats and oils. Its thermodynamic properties and kinetics of thermal decomposition have been studied over a range of temperature. A study on the zeolite supported platinum catalyzed isomerization and aromatization of n-hexane has been reported. Its removal from hexane contaminated air streams using vapor phase biological reactor (VPBR) immobilized with hexane-degrading Aspergillus niger strain has been studied. The metabolism of n-hexane forms 2,5-hexanedione (2,5-HD), which is reported to induce neurotoxicity.

Application

Hexane may be used in the following studies:
  • Preparation of graft-copolymer chains of poly-(dimethylsiloxane)-g-poly(methyl methacrylate).
  • GC/MS characterization of essential oils isolated from various origins.
  • As solvent for the preparation of samples and standards during chromatographic studies.
  • To extract free and total gossypol in combination with acetone from cottonseed flakes.
Suitable for HPLC, spectrophotometry, environmental testing

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Covalent binding of a neurotoxic n-hexane metabolite: conversion of primary amines to substituted pyrrole adducts by 2,5-hexanedione.
A P DeCaprio et al.
Toxicology and applied pharmacology, 65(3), 440-450 (1982-09-30)
Cottonseed extraction with mixtures of acetone and hexane.
Kuk MS, et al.
Journal of the American Oil Chemists' Society, 82(8), 609-612 (2005)
Thermodynamic properties of n-hexane.
Grigoryev BA, et al.
International Journal of Thermophysics, 9(3), 439-452 (1988)
Reforming of hexane with Pt/zeolite catalysts.
Dai LX, et al.
Catalysis Letters, 45(1-2), 107-112 (1997)
The thermal decomposition of n-hexane.
Ebert KH, et al.
International Journal of Chemical Kinetics, 15(5), 475-502 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service