Skip to Content
Merck
All Photos(2)

Key Documents

1.04371

Supelco

Hexane

≥98% (GC), suitable for gas chromatography (GC) ECD and FID, SupraSolv®

Synonym(s):

Hexane, n-Hexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH3
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-777-6
NACRES:
NA.03
technique(s):
gas chromatography (GC): suitable
bp:
69 °C (lit.)
vapor pressure:
256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

Product Name

n-Hexane, for gas chromatography ECD and FID SupraSolv®

vapor density

~3 (vs air)

Quality Level

vapor pressure

256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

product line

SupraSolv®

Assay

≥98% (GC)

form

liquid

autoignition temp.

453 °F

potency

25000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

7.7 %

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.375 (lit.)

kinematic viscosity

0.50 cSt(20 °C)

bp

69 °C (lit.)

mp

−95 °C (lit.)

transition temp

flash point -22 °C

solubility

0.0095 g/L

density

0.659 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CCCCCC

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector).
SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimum signal-to-noise ratio leading to reliable and reproducible analysis results.

Application

Ideal for GC applications including sensitive detection in residue and environmental analysis.
The product is used as a solvent/reagent in:
  • sample preparation for determination of chlorinated pesticides according to EPA method 508
  • pesticide residue analysis by fast GC and GC-MS
  • determination of folpet and phthalimide in food by LC-MS
  • quantification of cyclic volatile methylsiloxanes in human blood by large volume injection coupled to GC/MS
  • detection of fatty acids in cooking oils by GC

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Artifact-controlled quantification of folpet and phthalimide in food by liquid chromatography-high resolution mass spectrometry
Badoud F, et al.
Food Control, 91, 412-420 (2018)
Fast GC, GC-MS and sample preparation methods for pesticide residue analysis in non-fatty baby food
Hercegova A and Matisova E
Acta Chimica Slovaca, 1(1), 112-123 (2008)
Effects of Repeated Heating on Fatty Acid Composition of Plant-Based Cooking Oils
Szabo Z, et al.
Foods (Basel, Switzerland), 11(2), 192-192 (2022)
Cyclic volatile methylsiloxanes in human blood as markers for ruptured silicone gel-filled breast implants
Rosendahl P, et al.
Analytical and Bioanalytical Chemistry, 408(12), 3309-3317 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service