Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hexane
Synonym(s):
n
-Hexane, Hexane
Linear Formula:
CH
3
(CH
2
)
4
CH
3
CAS Number:
110-54-3
Molecular Weight:
86.18
EC Number:
203-777-6
Products
Product Category
solvents (16)
Brand
Sigma-Aldrich (8)
Supelco (11)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Reference Material Type
Special Grade
ACS reagent (2)
Puriss (2)
Puriss P.A. (2)
Anhydrous (1)
analytical standard (1)
certified reference material (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-19 of 19
Compare
Product Number
Description
Pricing
270504
Hexane,
≥95%, suitable for UV/Vis spectroscopy and HPLC, suitable for EPA ACB B21-02, EPA OTM-45, and EPA ACB B23-05b
Expand
View Pricing
139386
Hexane,
≥99%, suitable for GC/MS, suitable for EPA 1613, ReagentPlus
®
Expand
View Pricing
1.04391
Hexane,
≥98% (GC), suitable for HPLC, LiChrosolv
®
Expand
View Pricing
1.04367
Hexane,
≥99% (GC), ACS reagent, EMSURE
®
Expand
View Pricing
1.04371
Hexane,
≥98% (GC), suitable for gas chromatography (GC) ECD and FID, SupraSolv
®
Expand
View Pricing
208752
Hexane,
≥95%, Laboratory Reagent, suitable for EPA 1613
Expand
View Pricing
32293
Hexane,
≥99% (GC), ACS reagent, suitable for GC/MS, puriss. p.a., reag. Ph. Eur.
Expand
View Pricing
650552
Hexane,
≥95%, HPLC Plus, for residue analysis, suitable for gas chromatography (GC) and HPLC, suitable for EPA 1613, EPA ACB B21-02, EPA OTM-45, and EPA ACB B23-05b
Expand
View Pricing
1.04374
Hexane,
≥98.50% (hexane isomers + methylcyclopentane, GC), ≥96.0% (GC), ACS reagent, reag. Ph. Eur., EMSURE
®
Expand
View Pricing
1.00795
Hexane,
≥98% (GC), suitable for GC/MS, SupraSolv
®
Expand
View Pricing
1.04372
Hexane,
≥99% (GC), suitable for UV/Vis spectroscopy, Uvasol
®
Expand
View Pricing
1.04369
Hexane,
≥99% (GC), suitable for gas chromatography (GC), hypergrade for organic trace analysis SupraSolv
®
Expand
View Pricing
1.07023
Hexane,
≥98.5% (Σ hexane isomers + methylcyclopentane ), EMPARTA
®
ACS reagent
Expand
View Pricing
1.03701
Hexane,
≥99% (GC), suitable for LC/MS, LiChrosolv
®
Expand
View Pricing
5.89590
Hexane,
95%, anhydrous
Expand
View Pricing
52750
Hexane,
analytical standard
Expand
View Pricing
PHR1555
Hexane,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing
34859-M
Hexane,
≥97.0% (GC), suitable for HPLC
Expand
View Pricing
32293-M
Hexane,
≥99% (GC), ACS reagent, reag. Ph. Eur.
Expand
View Pricing