主要文件
27019
Supelco
带有 PTFE/硅胶隔垫的带孔螺纹盖
black polypropylene hole cap, thread for 15-425, PTFE/silicone, septum thickness 1.5 mm, for use with 7 mL vial, pkg of 100 ea
登录查看公司和协议定价
选择尺寸
100 PKG
$179.00
$179.00
请联系客服了解存货情况
获取大包装报价
选择尺寸
变更视图
100 PKG
$179.00
About This Item
UNSPSC代码:
41122001
$179.00
请联系客服了解存货情况
获取大包装报价
物料
PTFE/silicone
black polypropylene hole cap
特点
closure type black polypropylene hole cap
包装
pkg of 100 ea
隔板厚度
1.5 mm
配件
thread for 15-425
相容性
for use with 7 mL vial
正在寻找类似产品? 访问 产品对比指南
应用
It may also be used in automated solid-phase microextraction-gas chromatography-mass selective detection (AutoSPME-GC-MSD) analyses for the determination of sugar-amino acid reaction mechanisms and production of branched-chain alkyl-substituted pyrazines.[1]
历史批次信息供参考:
分析证书(COA)
访问文档库
An Automated Solid-Phase Microextraction-Gas Chromatography-Mass Selective Detection Approach for the Determination of Sugar-Amino Acid Reaction Mechanisms.
Coleman WM and Lawson SN.
Journal of Chromatographic Science, 37 (10), 383-387 (1999)