Merck
LC/MS Analysis of Ancymidol Enantiomers on Astec® CYCLOBOND I 2000 HP-RSP
LC/MS Analysis of Ancymidol Enantiomers on Astec® CYCLOBOND I 2000 HP-RSP application for HPLC
Materiały
Standardowe
Numer produktu
Opis
Cennik
Ancymidol
BioReagent, suitable for plant cell culture
kolumna analityczna
Numer produktu
Opis
Cennik
Astec® CYCLOBOND I 2000 HP-RSP Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
CYCLOBOND I 2000 HP-RSP, 25 cm x 4.6 mm I.D., 5 μm particles (24024AST)
mobile phase
[A] 5 mM ammonium acetate, pH 6.0; [B] acetonitrile; (80:20, A:B)
flow rate
1 mL/min
column temp.
35 °C
detector
MS (m/z = 257)