Merck
HPLC Analysis of Elastase on Discovery® BIO PolyMA-SCX, Ion Exchange
HPLC Analysis of Elastase on Discovery® BIO PolyMA-SCX, Ion Exchange application for HPLC
Materiały
Standardowe
Numer produktu
Opis
Cennik
Elastase from porcine pancreas
Type III, lyophilized powder, Protein 55-85 %, ≥4.0 units/mg protein
Elastase from porcine pancreas
lyophilized powder, suitable for cell culture
Elastase from porcine pancreas
Type IV, Protein 50-90 %, lyophilized powder, ≥4.0 units/mg protein (biuret)
Elastase from human leukocytes
lyophilized powder, ≥50 units/mg protein (Bradford)
Elastase from porcine pancreas
Type I, ≥4.0 units/mg protein
Elastase from porcine pancreas
powder, white, ≥10 U/mg
Elastase from porcine pancreas
lyophilized, powder, white, ~8 U/mg
CONDITIONS
column
Discovery BIO PolyMA-SCX, 5cm × 4.6mm, 5μm particles (59601-U)
mobile phase
[A] 20 mM sodium phosphate, pH 7.0; [B] 20 mM sodium phosphate, 0.5 M NaCl, pH 7.0
gradient
0 to 100% B in 30 min. (linear)
flow rate
0.5 mL/min
column temp.
25 °C
detector
UV, 280 nm
injection
10 μL
Opis
Komentarz do analizy
Discovery BIO PolyMA-SCX and PolyMA-WAX columns give sharp, efficient peaks for a wide variety of proteins.