Kluczowe dokumenty
7750999
RheBuild® Kit
for use with 7750 TPMV Series
About This Item
Kod UNSPSC:
41115854
kompatybilność
for use with 7750 TPMV Series
Powiązane kategorie
Opis ogólny
These kits include all of the tools and parts you need to repair your Rheodyne injector. The parts included are those most likely to be damaged, worn, or lost. Kits for front-loading injectors include: rotor seal, stator face assembly, isolation seal, needle guide, needle port cleaner, 2 hex keys, operating instructions, and a mini-manual. Parts may vary, depending on valve model.
Polecane produkty
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Informacje prawne
RheBuild is a registered trademark of IDEX Health & Science, LLC
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)