Key Documents
Z332496
BRAND® flexible discharge tube for Dispensette®
for 1, 2, 5 and 10 mL Dispensette
About This Item
Kod UNSPSC:
41121500
producent / nazwa handlowa
BRAND
Długość
80 cm
Opis ogólny
PTFE, coiled with safety handle
Inne uwagi
BR707925 (India)
Informacje prawne
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG