Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 1810,00 zł |
Informacje o tej pozycji
1810,00 zł
feature
incremental 0.02 mL, DE-M marking, with recirculation valve
manufacturer/tradename
BRAND 4630331
volume range
0.5-5 mL
fitting
(GL 28/S 28), GL 32, GL 38, S 40
General description
Features and Benefits
- Extremely easy filling and dispensing
- Simple priming by optimized flow channels
- Rapid and safe volume adjustment with internal scalloped track
Other Notes
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description volume range 0.5-5 mL, with recirculation valve | description volume range 5-50 mL, with recirculation valve | description volume range 1-10 mL, with recirculation valve | description volume range 2.5-25 mL, with recirculation valve |
| manufacturer/tradename BRAND 4630331 | manufacturer/tradename BRAND 4630361 | manufacturer/tradename BRAND 4630341 | manufacturer/tradename BRAND 4630351 |
| feature incremental 0.02 mL, with recirculation valve, DE-M marking | feature incremental 0.2 mL, DE-M marking, with recirculation valve | feature incremental 0.05 mL, DE-M marking, with recirculation valve | feature incremental 0.1 mL, DE-M marking, with recirculation valve |
| fitting (GL 28/S 28), GL 38, GL 32, S 40 | fitting (GL 32-33), GL 38, S 40 | fitting (GL 28/S 28), GL 32, GL 38, S 40 | fitting (GL 32-33), GL 38, S 40 |
| volume range 0.5-5 mL | volume range 5-50 mL | volume range 1-10 mL | volume range 2.5-25 mL |
Still not finding the right product?
Explore all of our products under BRAND® Dispensette® S Organic, Digital bottle-top dispenser
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?