Kluczowe dokumenty
J4705
Sigma-Aldrich
Jatrorrhizine
from Tinospora sagittata, ≥95% (HPLC)
Synonim(y):
7,8,13,13α-Tetradehydro-3-hydroxy-2,9,10-trimethoxyberbinium, Neprotin, Yatrorizine
About This Item
Wzór empiryczny (zapis Hilla):
C20H20NO4
Numer CAS:
Masa cząsteczkowa:
338.38
Numer WE:
Kod UNSPSC:
12352200
pochodzenie biologiczne
Tinospora sagittata
Próba
≥95% (HPLC)
temp. przechowywania
2-8°C
InChI
1S/C20H19NO4/c1-23-18-5-4-12-8-16-14-10-19(24-2)17(22)9-13(14)6-7-21(16)11-15(12)20(18)25-3/h4-5,8-11H,6-7H2,1-3H3/p+1
Klucz InChI
MXTLAHSTUOXGQF-UHFFFAOYSA-O
Działania biochem./fizjol.
Alkaloid fitochemiczny występujący w roślinach leczniczych. Działa przeciwzapalnie. Potencjalny środek terapeutyczny w zaburzeniach żołądkowo-jelitowych.
Piktogramy
Environment
Hasło ostrzegawcze
Warning
Zwroty wskazujące rodzaj zagrożenia
Zwroty wskazujące środki ostrożności
Klasyfikacja zagrożeń
Aquatic Acute 1
Kod klasy składowania
13 - Non Combustible Solids
Klasa zagrożenia wodnego (WGK)
WGK 3
Not applicable
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Odwiedź Bibliotekę dokumentów
Hua-dong Zhu et al.
Journal of chromatography. A, 1218(34), 5867-5871 (2011-07-26)
This paper for the first time describes the development of micelle to solvent stacking (MSS) to nonaqueous capillary electrophoresis (NACE). In this proposed MSS-NACE, sodium dodecyl sulfate (SDS) micelles transport, release, and focus analytes from the sample solution to the
Beibei Zhang et al.
The Journal of pharmacy and pharmacology, 64(3), 413-419 (2012-02-09)
Postoperative ileus is major cause of postoperative complication and prolonged hospitalization. Jatrorrhizine, which is a protoberberine alkaloid isolated from the medicinal plants Berberis aristata and Coptis chinensis, has been found to increase contractility of gastric antral and ileum smooth muscles
Fengqing Liu et al.
Journal of pharmaceutical and biomedical analysis, 56(5), 1006-1015 (2011-09-06)
Incurred rabbit plasmas samples were utilized for method quality assessment in this study, where an optimized protein precipitation method for the preparation of rabbit plasma samples and a rapid and sensitive liquid chromatography-electrospray ionization-mass spectrometry for the simultaneous determination of
Rong Shi et al.
Biopharmaceutics & drug disposition, 33(3), 135-145 (2012-02-22)
Jatrorrhizine, a protoberberine alkaloid derived from Coptis chinensis, is currently under investigation as a natural gastric prokinetic drug candidate. In vitro and in vivo studies were conducted to characterize its pharmacokinetics and metabolism. After intravenous administration, the plasma concentration kinetics
Hong Lin Zhai et al.
Analytica chimica acta, 657(2), 131-135 (2009-12-17)
High-performance liquid chromatography (HPLC) fingerprint has been commonly used in the quality control and assessment of herbal medicines, and two-dimensional (2D) fingerprint obtained by means of HPLC-diode array detector (HPLC/DAD) can provide higher reliability. In this paper, an approach to