Merck
Z146811
Sigma-Aldrich
Hose adapter
brass
About This Item
Kod UNSPSC:
41121815
NACRES:
NB.51
Materiały
brass
śr. wewn. węża
1/4 in.
dopasowywanie
1/4 in. female
Opis ogólny
Brass has serrated hose connector; SS has tapered hose connector.