Merck
HS120686
Clinical 4-Way Tube Rack
green
ログイン組織・契約価格を表示する
サイズを選択してください
5 EACH
¥12,990
¥12,990
Check Cart for Availability
サイズを選択してください
表示を変更する
5 EACH
¥12,990
About This Item
UNSPSCコード:
41122800
NACRES:
NB.17
¥12,990
Check Cart for Availability
物質
green
polypropylene
特徴
to hold Side A 12 x 13 mm, Side B 18 x 13 mm, Side C 12 x 17 mm and side D 18 x 17 mm tubes
包装
pack of 5 ea
メーカー/製品名
Heathrow Scientific 120686
L × W × H
232 mm × 70 mm × 95 mm
類似した製品をお探しですか? 訪問 製品比較ガイド
関連するカテゴリー
詳細
The Clinical 4-Way Tube Rack is the only tube rack specifically designed to hold round, flat & conical blood collection and culture tubes. It holds multiple tube sizes without multiple racks.
特徴および利点
  • Accommodates all brands of 13 and 16 mm round, flat and conical bottom blood collection & culture tubes
  • Only 4-way rack available with a six way connection, link different or the same well sizes, end to end or side by side, in any orientation
  • Customize the rack to fit the number of wells required, infinite units can be linked together or use one standalone unit
  • Uniformly spaced wells help eliminate pipetting errors which enables more accurate results
  • Molded alphanumeric identification for quick location and retrieval of tubes
  • Robust, made from polypropylene that is chemical resistant and easy to clean
  • Racks can be used in water baths and placed in freezers
  • Assorted color pack available for color coding options
  • Patent Pending
  • Autoclavable
最新バージョンのいずれかを選択してください:
試験成績書(COA)