Merck
HS120585
Magnetic Induction Stirrer
AC/DC input 100 V / 240 V, universal plug set, gray-blue
ログイン組織・契約価格を表示する
サイズを選択してください
1 EACH
¥65,570
¥65,570
Check Cart for Availability
サイズを選択してください
表示を変更する
1 EACH
¥65,570
About This Item
UNSPSCコード:
41103800
NACRES:
NB.72
¥65,570
Check Cart for Availability
物質
gray-blue
silicone plate
包装
pkg of 1 ea
メーカー/製品名
Heathrow Scientific 120585
パラメーター
2,000 rpm speed (max.)
プレート直径
160 mm
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
Strong, motor-less and maintenance free
Superior Functional Design
  • Advanced magnetic coil technology drives the spinning of the stir bar maintaining continuous, constant speed for reproducible results
  • Magnetic coupling is optimized by using alternating current (AC) as its driving force strong enough to stir viscous solutions
  • Motor-less operation generates minimal heat ideal for stirring temperature sensitive samples and environments for long periods
  • With no moving parts, produces less vibration and quieter operation over traditional magnetic stirrers
Intuitive Operation
  • Multiple stirring modes are easy to program
  • Bright digital display precisely controls time and speed
  • Quickly meets stirring speeds with rapid acceleration and braking (less than 10 seconds)
包装
INCLUDES:
Magnetic Induction Stirrer
2 Stir Bars 35 mm x 12 mm, Cylindrical Shape
1 Silicone Anti-skid Plate Cover
1 Low Voltage, Double Insulated Power Adapter (110-240 VAC, 50/60 Hz) with 4 Interchangeable Plugs
最新バージョンのいずれかを選択してください:
試験成績書(COA)