Documenti fondamentali
Z688371
Tough-Spots®
size diam. 1/2 in, white, pkg of 1000 roll
Sinonimo/i:
Tough-Spots®, solvent resistant labels, temperature resistant labels
Scegli un formato
1 EA
31,60 €
31,60 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
1 EA
31,60 €
About This Item
Codice UNSPSC:
55121611
NACRES:
NB.35
31,60 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
polypropylene
white
Confezionamento
pkg of 1000 roll
Produttore/marchio commerciale
Diversified Biotech T-SPOTS-50
Categorie correlate
Descrizione generale
Tough - Chemically inert, polyolefin labels resist most organic solvents and caustic agents. They adhere to most plastics†, glass, and metals without cracking, peeling, or degrading
Temperature-Resistant - From -196°C to 100°C, Tough-Spots® withstand boiling water baths, moderate oven temperatures (150°C), autoclaving and extreme freezer temperatures (liquid and vapor phase nitrogen)
Specially Designed - Label sheets are specially top-coated to readily accept laser printing. Heat-resistant, stay-flat sheets will not jam in laser printers
Ideally Sized - Fits a variety of microcentrifuge tube tops, cryovial caps and lids, as well as many other laboratory container caps, lids and covers
Color Coding - 3/8 in. (9.5 mm) and 1/2 in. (13 mm) diameter Laser Tough-Spots® are available in White, Blue, Green, Orange, Red and Yellow
Circular cryogenic labels formatted for handwriting. Tough-Spots® on a roll are easy to write PVC cryogenic labels that accept writing from most lab markers.
Caratteristiche e vantaggi
  • Chemically inert, polyolefin labels resist most organic solvents and caustic agents.
  • They adhere to most plastics†, glass, and metals without cracking, peeling, or degrading
  • Temperature-Resistant from -196°C to 100°C
  • Withstand boiling water baths, moderate oven temperatures (150°C), autoclaving and extreme freezer temperatures (liquid and vapor phase nitrogen)
  • Label sheets are specially top-coated to readily accept laser printing. Heat-resistant, stay-flat sheets will not jam in laser printers
  • Fits a variety of microcentrifuge tube tops, cryovial caps and lids, as well as many other laboratory container caps, lids and covers
Note legali
Tough-Spots is a registered trademark of Diversified Biotech
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 10
1 of 10
Laser Tough-Spots® diam. 1/2 in., assorted colors, pack of 3840 (192 labels per sheet)
Z743583
Laser Tough-Spots®
Laser Tough-Spots® diam. 1/2 in., green
L6164
Laser Tough-Spots®
Laser Tough-Tags™ Microcentrifuge tube labels, white, L × W 0.94 in. × 0.5 in.
Z688363
Laser Tough-Tags
Tough-Spots® size 1/2 in., blue
Z740262
Tough-Spots®
Tough-Spots® size 1/2 in., red
Z740265
Tough-Spots®
Tough-Spots® size 3/8 in., blue
Z740256
Tough-Spots®
Tough-Spots® 5 across, diam. 1/2 in., assorted colors, roll of 5000
Z743588
Tough-Spots®