Documenti
Z740256
Tough-Spots®
size 3/8 in., blue
Sinonimo/i:
solvent resistant labels, temperature resistant labels
About This Item
Codice UNSPSC:
55121611
NACRES:
NB.35
Materiali
PVC
blue
Confezionamento
pkg of 1000 ea (per roll)
Produttore/marchio commerciale
Diversified Biotech T-SPOTS-B
Misura
3/8 in.
Categorie correlate
Descrizione generale
Tough - Chemically inert, polyolefin labels resist most organic solvents and caustic agents. They adhere to most plastics†, glass, and metals without cracking, peeling, or degrading
Temperature-Resistant - From -196°C to 100°C, Tough-Spots® withstand boiling water baths, moderate oven temperatures (150°C), autoclaving and extreme freezer temperatures (liquid and vapor phase nitrogen)
Specially Designed - Label sheets are specially top-coated to readily accept laser printing. Heat-resistant, stay-flat sheets will not jam in laser printers
Ideally Sized - Fits a variety of microcentrifuge tube tops, cryovial caps and lids, as well as many other laboratory container caps, lids and covers
Color Coding - 3/8 in. (9.5 mm) and 1/2 in. (13 mm) diameter Laser Tough-Spots® are available in White, Blue, Green, Orange, Red and Yellow
Note legali
Tough-Spots is a registered trademark of Diversified Biotech
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 10
1 of 10
Tough-Spots® diam. 3/8 in., lavender, roll of 1000
Z743606
Tough-Spots®
Tough-Spots® diam. 3/8 in., pink, roll of 1000
Z743609
Tough-Spots®
Tough-Spots® diam. 3/8 in., gold, roll of 1000
Z743605
Tough-Spots®
Tough-Spots® 5 across, diam. 3/8 in., assorted colors, roll of 5000
Z743601
Tough-Spots®
Tough-Spots® diam. 1/2 in., lavender, roll of 1000
Z743592
Tough-Spots®
Tough-Spots® size 1/2 in., red
Z740265
Tough-Spots®