Documenti fondamentali
TMO140000
Nunc® EasyFill Cell Factory System
1 layer, polystyrene, surface area 632 cm2, sterile, vented, case of 6 ea
Sinonimo/i:
closed cell system layered storage rack, closed cell system rack
Scegli un formato
6 EACH
609,00 €
609,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
6 EACH
609,00 €
About This Item
Codice UNSPSC:
41161502
NACRES:
NB.13
609,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
polystyrene
Sterilità
sterile
Caratteristiche
vented
Confezionamento
case of 6 ea
Produttore/marchio commerciale
Nunc 140000
Area superficiale
632 cm2
Tipo di binding
Nunclon Delta surface
Descrizione generale
The Nunc® EasyFill Cell Factory systems are a space saving, out-of-the-box, multilayer solution and are as easy to use as a T-flask. Scale up your research and commercial-scale applications with confidence. These systems come with a cerrtified Nunclon® Delta cell culture surface to enable consistent performance from lot to lot and format to format. The 10-layer, single-use system holds the equivalent of 36 T175 flasks.
Caratteristiche e vantaggi
  • "Out of the box" system with no accessories required.
  • Large, vented screw closure for easy handling
  • Versatile port design allows for pouring and aseptic filling.
  • Fill and empty five times faster compared to T175 flasks.
  • Enhances productivity by enabling faster operations.
  • Fast start-up with a simple pour of media and cells into the large opening.
  • Versatile port facilitates pouring and aseptic filling techniques.
Note legali
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunclon is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)