This product does not include tips. The manufacturer recommends polycarbonate tips with an OD of 0.34mm (340 µm) for use with this instrument. The volume range for each delivery is between 0.25µL and 3µL. The ID is not specified.
P7289
Drummond Sequencing Pipette
volume 0.05-3 μL , adjustable in 0.05 μL increment
Scegli un formato
726,00 €
Scegli un formato
About This Item
726,00 €
Prodotti consigliati
Caratteristiche
electronic: no
single-channel
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Drummond Scientific 3-000-203
Volume
0.05-3 μL , adjustable in 0.05 μL increment
Categorie correlate
Descrizione generale
Includes 6 tips (0.34 mm O.D.), two replacement O-rings, and one plastic spacer.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
-
What is the ID and OD of the tip? what is the material of the tip? Is the tip a tapped shape to avoid clog? I am transferring suspension liquid with particle size at 25um blended with water.
1 risposta-