Documenti fondamentali
GN650160
Greiner CELLSTAR® multiwell culture plates
96 well, TC treated, clear
Sinonimo/i:
culture dishes, culture plates, multi dishes, Greiner CELLSTAR® multiwell culture plates
About This Item
Codice UNSPSC:
41122107
Materiali:
polystyrene
Prezzi e disponibilità al momento non sono disponibili
Materiali
polystyrene
Sterilità
sterile
Confezionamento
pack of 100 ea
Produttore/marchio commerciale
Greiner 650160
Disponibilità
available only in China
Descrizione generale
The chimney shape design, in combination with the condensation rings in the lid, effectively reduces the risk of cross-contamination and reduces evaporation from the wells. All plates can be stacked on top of each other, both with and without lids. The special design of the plates enables rapid temperature exchange across the entire base area of ​​each individual plate, even when the plates are stacked on top of each other. The standardization of depth, diameter and arrangement of the wells simplifies manual and automated work, as well as the use of common membrane insert systems. All plates have a printed lot number and are both pyrogen-free and DNase/RNase-free.
Note legali
CELLSTAR is a registered trademark of Greiner Bio-One GmbH
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)