Documenti fondamentali
CLS9018
Corning® 96 Well EIA/RIA Assay Microplate
flat bottom clear, polystyrene, high binding surface, bag of 25, non-sterile, lid: no
Sinonimo/i:
96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates
Scegli un formato
100 EA
294,00 €
294,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
100 EA
294,00 €
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.13
294,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
clear bottom
flat bottom clear
polystyrene
polystyrene
Sterilità
non-sterile
Caratteristiche
lid: no
skirt
Confezionamento
case of 100
bag of 25
Produttore/marchio commerciale
Corning 9018
Misura
96 wells
96 wells , 96 well plate
Volume pozzetto
330-360 μL , Note: CLS3366 and CLS3797 have well volume capacities of 330 uL
Volume di lavoro pozzetto
75-200 μL
Compatibilità
suitable for (EIA/RIA; immunoassays)
Tipo di binding
high binding surface
Descrizione generale
  • High binding surface is capable of binding medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions
  • Binding capacity is approximately 400 to 500 ng IgG/cm2
  • Ideal for immunoassays
  • Without lids, lids supplied separately
  • Total volume is 360 μL; recommended working volume of 75 to 200 μL
  • Conforms to standard microplate footprint and dimensions
These 96 well EIA/RIA plates are designed for in vitro diagnostic immunoassays. All EIA/RIA plates are manufactured from special medical-grade polystyrene for uniform binding, high optical clarity and low background absorption. In addition, the Easy-Wash products are part of the Certified Surface Chemistry Programme, which guarantees protein binding performance to be within QA release specifications.
Note legali
Corning is a registered trademark of Corning, Inc.
Easy-Wash is a trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)