Documenti fondamentali
CLS3073
Corning® CellBIND® Surface cell culture flasks
CellBIND 100cm2 canted neck low profile flask w/ Vent Cap, PS, sterile, 60/cs
Sinonimo/i:
cell culture flasks, tissue culture flasks
About This Item
Codice UNSPSC:
41122104
NACRES:
NB.13
Materiali
angled neck flask (low profile)
cap (vented)
high-density polyethylene cap
rectangular bottom flask
rectangular flask
rectangular polystyrene
vent cap
Sterilità
sterile; γ-irradiated
Caratteristiche
cap
graduations
Confezionamento
pack of 6
case of 60
Produttore/marchio commerciale
Corning 3073
Volume massimo
30 mL
Area superficiale
100 cm2
Volume d’esercizio
20-30 mL
Compatibilità
suitable for (cell culture applications; optimal for adherent cell culturing)
Tipo di binding
CellBIND® surface
Descrizione generale
The Corning CellBIND surface is produced by a microwave process which makes a more hydrophilic surface giving more consistent, even cell attachment, increased cell growth and yields. CellBIND enhances cell attachment and growth under difficult conditions, including reduced serum or serum-free medium.
CellBIND may provide a more ecominic alternative to expensive and unstable biological coatings.
Both enzymatic and non-enzymatic dissociating solutions have been used to remove and harvest cells from CellBIND surfaces, scraping is not recommended due to the increased strength of cell attachment.
  • Manufactured from optically clear virgin polystyrene.
  • Treated CellBIND surface treatment for enhanced cell attachment
  • 100% integrity tested.
  • Sterilized by gamma irradiation and certified nonpyrogenic
Note legali
CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)