Documenti fondamentali
212288
Sigma-Aldrich
Disperse Orange 1
Dye content ~15 %
Sinonimo/i:
4-(4-Nitrophenylazo)diphenylamine
About This Item
Formula condensata:
O2NC6H4N=NC6H4NHC6H5
Numero CAS:
Peso molecolare:
318.33
Colour Index:
11080
Numero CE:
Numero MDL:
Codice UNSPSC:
12171500
ID PubChem:
NACRES:
NA.47
Stato
powder
Composizione
Dye content, ~15%
λmax
483 nm
applicazioni
diagnostic assay manufacturing
hematology
histology
Temperatura di conservazione
room temp
Stringa SMILE
[O-][N+](=O)c1ccc(cc1)\N=N\c2ccc(Nc3ccccc3)cc2
InChI
1S/C18H14N4O2/c23-22(24)18-12-10-17(11-13-18)21-20-16-8-6-15(7-9-16)19-14-4-2-1-3-5-14/h1-13,19H/b21-20+
YFVXLROHJBSEDW-QZQOTICOSA-N
Codice della classe di stoccaggio
13 - Non Combustible Solids
WGK 3
Not applicable
Dispositivi di protezione individuale
Eyeshields, Gloves, type N95 (US)
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Late patch test reaction to Disperse Orange 1 not related to active sensitization.
L Malinauskiene et al.
Contact dermatitis, 63(5), 298-299 (2010-10-16)
Gareth O S Williams et al.
Lab on a chip, 12(18), 3356-3361 (2012-07-07)
Photonic crystal fibre constitutes an optofluidic system in which light can be efficiently coupled into a solution-phase sample, contained within the hollow core of the fibre, over long path-lengths. This provides an ideal arrangement for the highly sensitive monitoring of
Elisa R A Ferraz et al.
Journal of toxicology and environmental health. Part A, 75(16-17), 991-999 (2012-08-03)
During the dyeing process in baths approximately 10 to 15% of the dyes used are lost and reach industrial effluents, thus polluting the environment. Studies showed that some classes of dyes, mainly azo dyes and their by-products, exert adverse effects
G A R Oliveira et al.
Mutation research, 703(2), 200-208 (2010-09-21)
The treatment of textile effluents by the conventional method based on activated sludge followed by a chlorination step is not usually an effective method to remove azo dyes, and can generate products more mutagenic than the untreated dyes. The present
Laura Malinauskiene et al.
Dermatitis : contact, atopic, occupational, drug, 22(6), 335-343 (2012-06-02)
It is known that some patch-test preparations containing disperse dyes contain impurities with unknown relevance for the development or elicitation of contact allergy. To evaluate the significance of the impurities found in the commercial dyes Disperse Orange 1 (DO1) and