Documenti
CLS9999915
Corning® disposable phenolic cap
15-415 thread
Sinonimo/i:
phenolic caps, threaded cap, threaded caps
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.43
Produttore/marchio commerciale
Corning 99999-15
Altezza
16.0 mm
Raccordi
15-415 thread
Descrizione generale
These disposable Corning caps are made from phenolic resin and are autoclavable at 121°C. Caps are provided with a white rubber liner.
Note legali
Corning is a registered trademark of Corning, Inc.