Documenti fondamentali
BR782305
BRAND® strip plates
size 96 wells, 12xF8, HB,
dividable: no
Sinonimo/i:
BRANDplates®, Microplates, Plates, Strip Plates
Scegli un formato
100 EA
480,00 €
480,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Scegli un formato
Cambia visualizzazione
100 EA
480,00 €
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.22
Materiali:
polystyrene
transparent wells
Misura:
96 wells
Tipo di binding:
high binding surface
Caratteristiche:
12xF8, HB
immunoGrade
480,00 €
Per informazioni sulla disponibilità, contatta il Servizio Clienti.
Materiali
polystyrene
transparent wells
Caratteristiche
12xF8, HB
immunoGrade
Confezionamento
pack of 100 ea
pkg of (4 bags of 25)
Produttore/marchio commerciale
BRAND 782305
Misura
96 wells
Tipo di binding
high binding surface
Descrizione generale
BRANDplates® microtitration plates offer ideal visual properties and are produced according to ANSI/SLAS standards. They offer excellent flexibility. Only as many cavities as necessary are used. Two versions are available :Frame without grate:for processing 12 strips of 8 wells. Individual strips can be removed from the frame and re-inserted. Strips are not separable.Frame with grate :also 12 strips of 8 wells. Individual strips can be removed from and re-inserted into the plate. Strips can be divided in up to 8 individual tubes and inserted individually into the frame plate.They are available as 96-well standard plates as strip plates with immunoGrade surface – for greater flexibility and optimal results.immunoGrade high binding capacity (high-binding), ideal for IgG and molecules with hydrophobic and hydrophilic regions.CERTIFIED LIFE SCIENCE QUALITY: Virgin raw materials, Manufactured under controlled room conditions, Intensive in-process controls, Visual inspections, Batch management, Final product inspection, Proven functionality
Caratteristiche e vantaggi
• Optimal surfaces for optimal results• Reliable quality from the cleanroom• Tested by independent laboratories
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)