Documenti fondamentali
BR143260
BRAND® special laboratory tubing
I.D. × O.D. × W 4 mm × 6 mm × 1 mm, L 20 m
Sinonimo/i:
Special laboratory tubing, Tubing
About This Item
Codice UNSPSC:
41105108
NACRES:
NB.35
Materiali
PVC
Produttore/marchio commerciale
BRAND 143260
Diam. int. × diam. est. × largh.
4 mm × 6 mm × 1 mm
Lunghezza
20 m
Descrizione generale
Particularly high-grade material, manufactured to a first-class standard. Good flexibility and chemical resistance make this tubing ideal for use with apparatus assemblies. Autoclavable (121 °C).
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)