Ugrás a tartalomra
Merck
  • Influence of Dispersing Method on the Quality of Nano-Admixtures Homogenization in Cement Matrix.
Influence of Dispersing Method on the Quality of Nano-Admixtures Homogenization in Cement Matrix.
Materials (Basel, Switzerland) (2020-11-05)
Elżbieta Horszczaruk, Paweł Łukowski, Cyprian Seul
KIVONAT
In recent years, a nano-modification of the cement composites allowed to develop a number of new materials. The use of even small amount of nano-admixture makes possible not only to improve the physico-mechanical properties of the cement materials, but also to obtain the composite with high usability, optimised for the given application. The basic problem of nano-modification of the cement composites remains the effectiveness of dispersing the nanomaterials inside the cement matrix. This paper deals with the effect of the type and size of the nanoparticles on the tendency to their agglomeration in the cement matrix. The main techniques and methods of dispersing the nanomaterials are presented. It has been demonstrated, on the basis of the results of testing of three nanomodifiers of 0D type (nano-SiO2, nano-Fe3O4 and nano-Pb3O4), how the structure and properties of the nanomaterial affect the behaviour of the particles when dissolving in the mixing water and applying a superplasticiser. The nanoparticles had similar size of about 100 nm but different physico-chemical properties. The methods of dispersing covered the use of high-speed mechanical stirring and ultrasonication. The influence of the method of nano-modifier dispersing on the mechanical performance of the cement composite has been presented on the basis of the results of testing the cement mortars modified with 3% admixture of nano-SiO2.
ANYAGOK
Cikkszám
Márka
Termékleírás
Sigma-Aldrich
Iron(II,III) oxide, nanopowder, 50-100 nm particle size (SEM), 97% trace metals basis
Sigma-Aldrich
Lead oxide, powder (red), 1-2 μm, 99%