Fontos dokumentumok
Z765570
Tube rack for Benchmixer XL multi-tube vortexer
Holds 50 x 13 mm tubes
About This Item
UNSPSC kód:
41122804
NACRES:
NB.77
Jellemzők
Holds 50 x 13 mm tubes
kiszerelés
pkg of 1 ea
gyártó/kereskedő neve
Benchmark Scientific BV1010-13
Általános leírás
This rack enables use of 50 x 13 mm tubes in the BenchMixer XL heavy-duty hands-free mixing vortex mixer. Ideal for the QuEChERS pesticide analysis method.
Benchmixer XL vortexer head adapters to hold multiple tubes.
Jogi információk
Benchmixer is a trademark of Benchmark Scientific, Inc.