Fontos dokumentumok
GE28-9323-63
Vivaspin® 20, 100 kDa MWCO Polyethersulfone
Cytiva 28-9323-63, pack of 12 pieces
About This Item
UNSPSC kód:
41104916
NACRES:
NB.24
material:
polycarbonate housing
polyethersulfone membrane
pórusméret:
100 kDa MWCO
csőméret:
50 mL , conical bottom
hasznos térfogat:
20 mL
sterilitás:
non-sterile
kiszerelés:
pack of 12 pieces
material
polycarbonate housing
polyethersulfone membrane
sterilitás
non-sterile
kiszerelés
pack of 12 pieces
gyártó/kereskedő neve
Cytiva 28-9323-63
elérhetőség
not available in Japan
csőméret
50 mL , conical bottom
hasznos térfogat
20 mL
pórusméret
100 kDa MWCO
tárolási hőmérséklet
room temp
Általános leírás
Designed for fast, nondenaturing concentration of biological samples by membrane ultrafiltration. Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%.
Vivaspin® sample concentrators are designed for fast, nondenaturing concentration of biological samples by membrane ultrafiltration. Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. The entire process is performed in a single tube with an upper compartment containing sample and lower compartment separated by a semipermeable membrane with a molecular weight cut-off (MWCO) selected by the user. Centrifugation is applied to force solvent through the membrane, leaving a more concentrated sample in the upper chamber.
Vivaspin® sample concentrators cater for sample volumes from 100 μl to 20 ml, with a range of molecular weight cutoff values from Mr 3 000 to 100 000. For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest
Tulajdonságok és előnyök
  • Compatible pH range is from pH 1 to 9.
  • Patented dead-stop technology, which ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery
  • One-step sample concentration in a single tube for minimal sample handling and reduced sample loss.
  • Vertical polyethersulfone membrane minimizes membrane blockage and tolerates high flow rates.
Tárolás és stabilitás
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Analízis megjegyzés
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Jogi információk
Vivaspin is a registered trademark of Sartorius Stedim Biotech GmbH
Tárolási osztály kódja
13 - Non Combustible Solids
Válasszon a legfrissebb verziók közül:
Analitikai tanúsítványok (COA)
Dokumentumtár megtekintése
Cikkek
This page describes troubleshooting strategies in purification of GST-tagged proteins using Cytiva products.
Protocols
Rules and good practice in sample preparation for Western blot sample preparation from cell culture and tissue samples.
This page shows how to perform sample desalting, buffer exchange and concentration for affinity chromatography of tagged proteins.