Fontos dokumentumok
ZSQ240GVT0
Milli-Q
Milli-Q® SQ Gravity Valve
Gravity dispensing of pure RO water from the Milli-Q® SQ Switch 3.5 L Tank
About This Item
UNSPSC kód:
41104200
eCl@ss:
33050190
NACRES:
NC.75
kiszerelés
pkg of 1 unit
parameter
6 L/min distribution flow rate (Pure RO water from SQ Gravity valve)
kompatibilitás
for use with Milli-Q® SQ 2Series
Általános leírás
To learn how Milli-Q® SQ 2Series water systems work and read a full overview, visit our dedicated page.
The Milli-Q® SQ Gravity Valve allows the dispensing of pure RO water from the Milli-Q® SQ Switch 3.5 L Tank.
It is a manual valve with 2 m long tubing that is connected to the back of the SQ Switch 3.5 L Tank base which is part of the Milli-Q® SQ 2Series family of water purification systems.
It includes a special key that facilitates tube disconnection.
Jogi információk
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Dokumentumtár megtekintése
Protocols
View instructional videos and download User Manuals for Milli-Q® SQ 2Series lab water systems. You’ll be up and running in no time. Buy online systems, accessories, cartridges and services.
Related Content
Discover a new way to think about ultrapure water. Now any lab bench with a plug can freshly dispense 18.2 MΩ∙cm ultrapure water. Robust Quality. Simple. Compact. Scalable. A Greener Alternative Product. Buy Online.