Fontos dokumentumok
ZLXLCLPAK
Milli-Q
RO Regeneration Tool
For use with Milli-Q® CLX/HX/HR/HXSD 7000 series water purification systems
About This Item
UNSPSC kód:
41104200
NACRES:
JA.13
kiszerelés
pkg of 1 unit
kompatibilitás
for use with Milli-Q® CLX
for use with Milli-Q® HR
for use with Milli-Q® HX
Általános leírás
The RO Regeneration Tool is a water purification accessory used with high-volume water systems with poor feed water conditions requiring additional maintenance of the reverse osmosis (RO) membrane.
Alkalmazás
For high volume water systems requiring additional RO membrane maintenance with poor feed water conditions.
Jogi információk
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany