跳转至内容
Merck
  • Obif, a Transmembrane Protein, Is Required for Bone Mineralization and Spermatogenesis in Mice.
Obif, a Transmembrane Protein, Is Required for Bone Mineralization and Spermatogenesis in Mice.
PloS one (2015-07-25)
Koji Mizuhashi, Taro Chaya, Takashi Kanamoto, Yoshihiro Omori, Takahisa Furukawa
摘要
Various kinds of transmembrane and secreted proteins play pivotal roles in development through cell-cell communication. We previously reported that Obif (Osteoblast induction factor, Tmem119), encoding a single transmembrane protein, is expressed in differentiating osteoblasts, and that Obif-/- mice exhibit significantly reduced bone volume in the femur. In the current study, we characterized the Obif protein and further investigated the biological phenotypes of a variety of tissues in Obif-/- mice. First, we found that O-glycosylation of the Obif protein occurs at serine residue 36 in the Obif extracellular domain. Next, we observed that Obif-/- mice exhibit bone dysplasia in association with significantly increased osteoid volume per osteoid surface (OV/OS) and osteoid maturation time (Omt), and significantly decreased mineral apposition rate (MAR) and bone formation rate per bone surface (BFR/BS). In addition, we observed that Obif-/- mice show a significant decrease in testis weight as well as in sperm number. By histological analysis, we found that Obif is expressed in spermatocytes and spermatids in the developing testis and that spermatogenesis is halted at the round spermatid stage in the Obif-/- testis that lacks sperm. However, the number of litters fathered by male mice was slightly reduced in Obif-/- mice compared with wild-type mice, although this was not statistically significant. Our results, taken together with previous observations, indicate that Obif is a type Ia transmembrane protein whose N-terminal region is O-glycosylated. In addition, we found that Obif is required for normal bone mineralization and late testicular differentiation in vivo. These findings suggest that Obif plays essential roles in the development of multiple tissues.
材料
货号
品牌
产品描述
Sigma-Aldrich
单克隆抗-FLAG® M2 小鼠抗, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
L -抗坏血酸, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L -抗坏血酸, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L -抗坏血酸, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
四环素 盐酸盐, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
胆钙化醇, ≥98% (HPLC)
Sigma-Aldrich
L -抗坏血酸, reagent grade, crystalline
Sigma-Aldrich
L -抗坏血酸, reagent grade
Sigma-Aldrich
L -抗坏血酸, meets USP testing specifications
Supelco
维生素D3标准液 CRM 溶液, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
L -抗坏血酸, FCC, FG
Sigma-Aldrich
L -抗坏血酸, 99%
Sigma-Aldrich
L -抗坏血酸, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Vitamin D3 solution, 100 μg/mL in ethanol, 97% (CP)
Sigma-Aldrich
L -抗坏血酸, ACS reagent, ≥99%
Sigma-Aldrich
四环素 盐酸盐, ≥95% (European Pharmacopoeia HPLC assay)
Sigma-Aldrich
L -抗坏血酸, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
四环素 盐酸盐, powder
Sigma-Aldrich
L -抗坏血酸, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
四环素 盐酸盐, meets USP testing specifications
Sigma-Aldrich
胆钙化醇, meets USP testing specifications
Sigma-Aldrich
胆钙化醇, analytical standard