GE29-0119-27
1 of 10
GERPN6302
碘代乙酰胺
GE17-6001-13
Immobiline® Drystrip pH 4-7, 13 cm
GE17-6003-18
DeStreak™ Reagent
Sigma-Aldrich
U5128
尿素
GE18-1016-60
Immobiline® Drystrip pH 4-7, 11 cm
Supelco
646563
DL-二硫代苏糖醇 溶液
43816
Millipore
324506
EDTA,0.5 M,pH 8.0,分子生物学级,DEPC 处理
M3148
2-巯基乙醇
M6250
pkg of 1 L
Cytiva 29011927
1 L
12 - Non Combustible Liquids
历史批次信息供参考:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 文件 section.