Merck
23240-U
Supelco
Schraubkappe
black phenolic solid cap (with polyseal liner), thread for 22-400, pkg of 144 ea
About This Item
UNSPSC-Code:
41122001
NACRES:
NB.11
Materialien
black phenolic solid cap (with polyseal liner)
Verpackung
pkg of 144 ea
Fitting
thread for 22-400
Kompatibilität
for use with 120 mL (49 × 114 mm)
Allgemeine Beschreibung
Screw caps (thread 22-400) are available for use with sealing septa and solid caps with polyseal liners. Caps are offered in solid phenolic caps that can withstand a maximum temperature of 205° C.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)