Merck
Z137235
Sigma-Aldrich
Ace round-bottom flask with threaded side-arm
capacity 1,000 mL
Synonym(e):
bottom flask, flask round, round bottom flasks
Größe auswählen
1 EA
125,00 €
125,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
125,00 €
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
125,00 €
Check Cart for Availability
Materialien
borosilicate glass
round bottom flask
short
two-neck
Sterilität
non-sterile
Leistungsmerkmale
closure type Standard Taper Joint
Verpackung
pack of 1
Hersteller/Markenname
Ace Glass 6936-44
Kapazität
1,000 mL
Allgemeine Beschreibung
24/40 joint. Side-arm is internally threaded for use with a nylon bushing. Will accommodate thermometers or bleed tubes up to 7 mm diam. Complete with bushing and FETFE® O-ring.
Rechtliche Hinweise
FETFE is a registered trademark of Ace Glass, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)