Hlavní dokumenty
56051
Tubing Coupler
1/4-28 female Omnifit union for 1/4-28 male nut
About This Item
UNSPSC Code:
12000000
material
Tefzel®
packaging
pkg of 10 ea
technique(s)
LPLC: suitable
fitting
1/4-28 female Omnifit union for 1/4-28 male nut
Legal Information
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.
Zákazníci si také prohlíželi
Slide 1 of 2
1 of 2
Union PEEK, for connecting 1/16 in., bore size 0.25 mm
Z227242
Union