Hlavní dokumenty
55073
Upchurch Tubing Adapter
About This Item
UNSPSC Code:
41115700
material
PEEK adapter (one-piece design)
packaging
pkg of 1 ea
fitting
male Luer (lock hub-quick connect)
10-32 female
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)