Key Documents
27215
Supelco
Crimp seals with PTFE/silicone septa
silver aluminum seal, seal diam. 11 mm, white PTFE/silicone septum
About This Item
UNSPSC Code:
41121821
material
silver aluminum seal
white PTFE/silicone septum
feature
closure type crimp seal
packaging
pkg of 1000 ea
seal diam.
11 mm
fitting
thread for 11 mm
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)