Key Documents
26252
SGE MicroVolume syringes with standard plunger, fixed needle
500F, volume 500 μL, needle size 25 ga, needle L 50 mm
Synonyma:
SYRINGE SGE 500UL FN PT#2,25GA
About This Item
UNSPSC Code:
41122001
eCl@ss:
32070500
material
bevel tip stainless steel needle (point style 2)
needle size
25 ga
feature
needle type Fixed
packaging
pkg of 1 ea
manufacturer/tradename
SGE 007000
needle L
50 mm
volume
500 μL
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)