Hlavní dokumenty
22780
Supelco
Fritless Purge Sampler Glassware
size 5 mL, Tekmar® 14-2336-024
About This Item
UNSPSC Code:
41115700
agency
suitable for EPA 601
feature
fritted: no
manufacturer/tradename
Tekmar® 14-2336-024
size
5 mL
application(s)
industrial hygiene
compatibility
for use with Tekmar® 2000/3000
Application
For use with Tekmar 2000/3000 & Velocity XPT Concentrators
Legal Information
Tekmar is a registered trademark of Tekmar Co.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.
Navštívit knihovnu dokumentů
Související obsah
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.