Hlavní dokumenty
20389
Tubing Reamer
About This Item
UNSPSC Code:
41121800
technique(s)
gas chromatography (GC): suitable
General description
This 5 in. hand tool does a fine job of opening and rounding tubing ends thet are cut. The tip is stainless steel for durability and inertness; and the handle is wood for comfort.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)