P4169
Pipette workstation
acrylic, with upper and lower rack
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Doporučené produkty
material
acrylic
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18961-0060
General description
Acrylic rack holds pipettes of a wide variety of sizes on either the upper rack (25 mm wide slots) or the lower rack (20 mm wide slots).
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)