Key Documents
Z162078
Sigma-Aldrich
Scienceware® pipette support/rest
polystyrene, pkg of 1 ea
About This Item
UNSPSC Code:
41122801
NACRES:
NB.01
material
polystyrene
feature
16 small or 7 large pipettes
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art F18940-0000
L × W × H
280 mm × 215 mm × 38 mm
General description
Includes an off-center support for short pipettes, syringes, etc.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP