Vybrat velikost
| Vám/Skladová položka | Dostupnost | Cena |
|---|---|---|
1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 1 560,00 Kč |
O této položce
1 560,00 Kč
material
acrylic , clear
feature
to hold 4 pipette(s)
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS206204
L × W × H
199 mm × 172 mm × 206.4 mm
General description
Features and Benefits
- 4-Place Pipette Holder (1 in (25mm) Upper Slot, 0.8 in (20mm) Lower Slot)
- Designed to hold most major pipette brands
1 of 1
Tato položka | |||
|---|---|---|---|
| material acrylic , clear | material acrylic , clear | material acrylic , blue | material acrylic , acrylonitrile-butadiene-styrene (ABS) , blue |
| feature to hold 4 pipette(s) | feature to hold 6 pipette(s) | feature to hold 6 pipette(s) | feature to hold (pipettes), to hold 6 pipette(s) |
| manufacturer/tradename Heathrow Scientific HS206204 | manufacturer/tradename Heathrow Scientific HS20620 | manufacturer/tradename Heathrow Scientific HS20613C | manufacturer/tradename Heathrow Scientific HS20613E |
| L × W × H 199 mm × 172 mm × 206.4 mm | L × W × H 295.3 mm × 172 mm × 203.2 mm | L × W × H 295.3 mm × 172 mm × 203.2 mm | L × W × H 295.3 mm × 172 mm × 203.2 mm |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
Still not finding the right product?
Explore all of our products under Pipette Stand
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?