Hlavní dokumenty
EP5702721000
Caps for Eppendorf® A-4-38 Rotor
aerosol-tight, aerosol-tight, for 5702/570R/5702RH, for 100 mL round bucket, for 100 mL round bucket
About This Item
UNSPSC Code:
41103913
NACRES:
NB.82
feature
aerosol-tight
for 100 mL round bucket
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5702721000 (EU)
technique(s)
centrifugation: suitable
compatibility
for use with 5702 R
for use with 5702 RH
for use with 5702
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)