Key Documents
EP5702719005
Eppendorf® Adapter for 85/100 mL Round Bucket
holds Vacutainer and Falcon tubes 2.6-7ml, pack of 2 ea
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
feature
holds Vacutainer and Falcon tubes 2.6-7ml
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5702719005
technique(s)
centrifugation: suitable
compatibility
for use with 100 mL round buckets
Legal Information
Eppendorf is a registered trademark of Eppendorf AG