Key Documents
EP5910700008
Caps for Eppendorf® S-4x400 Rotor
Aerosol-tight, pack of 4 ea, for Centrifuge 5910 R
About This Item
UNSPSC Code:
41103913
NACRES:
NB.82
feature
Aerosol-tight
for Centrifuge 5910 R
packaging
pack of 4 ea
manufacturer/tradename
Eppendorf® 5910700008
technique(s)
centrifugation: suitable
compatibility
for use with 5910 R
Legal Information
Eppendorf is a registered trademark of Eppendorf AG